missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%, Honeywell
Supplier: Honeywell Inc LP34719
Specifications
| Toluene | |
| 108-88-3 | |
| 0.867 g/cm3 | |
| 19 L | |
| Metal Can | |
| C7H8 | |
| UN1294 | |
| 635760 | |
| 5.2% in Water at 20°C | |
| CC1=CC=CC=C1 | |
| 92.14 | |
| CHEBI:17578 | |
| >99.5% |
| ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8% | |
| 100.0000% | |
| 110.6°C | |
| 4°C | |
| by GC Analysis | |
| MFCD00008512 | |
| 0.03% max. | |
| methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
| YXFVVABEGXRONW-UHFFFAOYSA-N | |
| toluene | |
| 1140 | |
| 92.14 | |
| ACS Reagent |
Chemical Identifiers
| 108-88-3 | |
| 92.14 | |
| YXFVVABEGXRONW-UHFFFAOYSA-N | |
| 1140 | |
| toluene |
| C7H8 | |
| MFCD00008512 | |
| methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
| CHEBI:17578 | |
| CC1=CC=CC=C1 |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene