missing translation for 'onlineSavingsMsg'
Learn More

Toluene, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.8%, Honeywell™

Catalog No. p-7112011
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001720 4 x 4 L Glass Bottle
6001721 4 x 4 L Poly-coated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001720 Supplier Honeywell Chemicals Supplier No. AH3474
Only null left

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Absorbance 0.1 AU max. at 300 nm, 0.025 AU max. at 325 nm
CAS Max % 100.0000%
Density 0.867 g/cm3
Boiling Point 110.6°C
Quantity 4 x 4 L
Flash Point 4°C
Packaging Glass Bottle
UN Number UN1294
Beilstein 635760
Water 0.03% max.
Solubility Information 5.2% in Water at 20°C
Formula Weight 92.14
Percent Purity >99.8%
Grade HPLC
Chemical Name or Material Toluene, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, >99.8%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene