missing translation for 'onlineSavingsMsg'
Learn More

Toluene, ACS reagent, ≥99.5%, Solstice

Catalog No. 60044995
Change view
Click to view available options
Quantity:
2.5 L
5 L
Packaging:
Aluminum Bottle
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044995 2.5 L Glass Bottle
60044999 5 L Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60044995 Supplier Honeywell Inc Supplier No. 1794182.5L

ACS reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -93°C (literature)
CAS Max % 100.0000%
Density 0.865 g/mL at 25 °C(lit.)
Quantity 2.5 L
Flash Point 4°C
Packaging Glass Bottle
Linear Formula C6H5CH3
UN Number UN1294
Beilstein 635760
Boiling Point 110.6°C
Formula Weight 92.14
Percent Purity >99.5%
Grade ACS
Chemical Name or Material Toluene, ACS reagent, ≥99.5%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene
EINECSNumber 203-625-9
Recommended Storage Room Temp