missing translation for 'onlineSavingsMsg'
Learn More

Toluene ACS, MilliporeSigma™

Catalog No. MTX0735P4
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Metal Can
Poly-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MTX0735P4 4 L Poly-coated Glass Bottle
MTX07353 20 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MTX0735P4 Supplier Technidata Supplier No. TX0735P4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Density 0.87g/cm3 at 20°C
Quantity 4 L
Flash Point 4.4°C (39.9°F), Closed cup
Ignition Point 480°C (896°F)
Odor Aromatic
Packaging Poly-coated Glass Bottle
Linear Formula C6H5CH3
UN Number UN 1294
Boiling Point 110.6°C
Solubility Information Insoluble in water
Formula Weight 92.14
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Toluene
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-625-9
Recommended Storage Room Temperature