Learn More
Toluene, ACS Grade, ≥ 99.5 %, LabChem™
Supplier: LabChem LC2617044
Specifications
| Toluene | |
| 108-88-3 | |
| 100 | |
| 870kg/m3 | |
| 4 x 4 L | |
| Amber Glass | |
| ≤0.001% | |
| C7H8 | |
| MFCD00008512 | |
| UN1294 | |
| methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
| CC1=CC=CC=C1 | |
| 92.14 | |
| CHEBI:17578 | |
| ≥99.5% |
| -95°C | |
| Colorless | |
| Carbon dioxide; Carbon monoxide | |
| 111°C | |
| Passes Test | |
| Liquid | |
| 870kg/m3 | |
| C7H8 | |
| 0.69mm2/s | |
| ≤0.03% | |
| YXFVVABEGXRONW-UHFFFAOYSA-N | |
| toluene | |
| 1140 | |
| 92.14 | |
| ACS |
Chemical Identifiers
| 108-88-3 | |
| 92.14 | |
| YXFVVABEGXRONW-UHFFFAOYSA-N | |
| 1140 | |
| toluene |
| C7H8 | |
| MFCD00008512 | |
| methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
| CHEBI:17578 | |
| CC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Danger
EINECSNumber : 203-625-9
Recommended Storage : Room Temperature