missing translation for 'onlineSavingsMsg'
Learn More

Toluene, 99.5% min., ACS, MilliporeSigma™

Catalog No. MTX07356
Change view
Click to view available options
Quantity:
500 mL
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MTX07356 500 mL
MTX073520 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MTX07356 Supplier Technidata Supplier No. TX07356

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Color Colorless
Quantity 500 mL
Physical Form Liquid
Assay Percent Range 99.5% min.
Formula Weight 92.14
Chemical Name or Material Toluene, For ACS analysis