missing translation for 'onlineSavingsMsg'
Learn More

(Toluene-3,4-dithiolato)zinc(II) 95.0+%, TCI America™

Catalog No. T02795G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02795G 5 g
T02791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T02795G Supplier TCI America Supplier No. T02795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29726-21-4
Molecular Formula C7H8S2Zn
Molecular Weight (g/mol) 221.641
MDL Number MFCD00005863
InChI Key SVSNSFKOIZDLHR-UHFFFAOYSA-N
Synonym Zinc Dithiol
PubChem CID 57518019
IUPAC Name 4-methylbenzene-1,2-dithiol;zinc
SMILES CC1=CC(=C(C=C1)S)S.[Zn]

Specifications

Color White
Quantity 5 g
Formula Weight 219.63
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Toluene-3,4-dithiolato)zinc(II)