missing translation for 'onlineSavingsMsg'
Learn More

Tolmetin sodium salt dihydrate, 98+%

Catalog No. AAJ6266314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6266314 25 g
AAJ6266303 1 g
AAJ6266306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6266314 Supplier Thermo Scientific Chemicals Supplier No. J6266314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An NSAID compound | CAS: 64490-92-2 | C15H18NNaO5 | 315.301 g/mol

Tolmetin Sodium is an anti-inflammatory, antirheumatic agent. It can inhibit aldo-keto reductases (e.g., IC50 = 2.39 μM for human aldose reductase), a family of enzymes that catalyze the NADPH-dependent reduction of carbonyl groups in a number of important steroid, metabolic, and prostanoid molecules. It is a beta-adrenergic blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tolmetin Sodium is an anti-inflammatory, antirheumatic agent. It can inhibit aldo-keto reductases (e.g., IC50 = 2.39 µM for human aldose reductase), a family of enzymes that catalyze the NADPH-dependent reduction of carbonyl groups in a number of important steroid, metabolic, and prostanoid molecules. It is a beta-adrenergic blocker.

Solubility
Soluble in water

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 64490-92-2
Molecular Formula C15H18NNaO5
Molecular Weight (g/mol) 315.301
MDL Number MFCD00150761
InChI Key QQILXENAYPUNEA-UHFFFAOYSA-M
Synonym tolmetin sodium dihydrate, sodium tolmetin dihydrate, tolumetin sodium dihydrate, unii-02n1tzf99f, sodium 2-1-methyl-5-4-methylbenzoyl-1h-pyrrol-2-yl acetate dihydrate, tolmetin sodium salt dihydrate, sodium 1-methyl-5-p-toluoylpyrrole-2-acetate dihydrate, tolectin tn, 1-methyl-5-p-toluoyl pyrrole-2 acetic acid
PubChem CID 23677829
ChEBI CHEBI:72014
IUPAC Name sodium;2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetate;dihydrate
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=C(N2C)CC(=O)[O-].O.O.[Na+]

Specifications

Color White
Quantity 25 g
Merck Index 14,9518
Solubility Information Soluble in water
Formula Weight 315.30 (279.27 Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Tolmetin sodium salt dihydrate
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 252-687-3
RTECSNumber UX9280000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only