accessibility menu, dialog, popup

UserName

H2OH2ONaCH3NOOOH3C

CAS RN 64490-92-2

IUPAC Name: sodium 2-[1-methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl]acetate dihydrate
Synonyms: sodium 2-[1-methyl-5-(4-methylbenzoyl)pyrrol-2-yl]acetate dihydrate
Molecular Weight (g/mol): 315.3
Molecular Formula: C15H18NNaO5
InChi Key: QQILXENAYPUNEA-UHFFFAOYSA-M
SMILES: O.O.[Na+].CN1C(CC([O-])=O)=CC=C1C(=O)C1=CC=C(C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results