missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolbutamide, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 64-77-7 |
| Physical Form | Neat |
| Linear Formula | C12H18N2O3 S |
| MDL Number | MFCD00027169 |
| Synonym | 1-Butyl-3-(4-methylphenylsulfonyl)urea; N-[(Butylamino)carbonyl]-4-methylbenzenesulfonamide |
| Type | Analytical Standard |
| Quantity | 25 g |
| Formula Weight | 270.35 |
| Grade | Analytical Standard |