missing translation for 'onlineSavingsMsg'
Learn More

α-Tocopherol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012305
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012305 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012305 Supplier Merck Emd Millipore Supplier No. PHR1031500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

α-Tocopherol is a powerful biological antioxidant and the most active form of vitamin E in humans.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 10191-41-0
Density 0.950 g/mL (at 20°C)
Flash Point 113°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C29H50O2
MDL Number MFCD00006848
Synonym (+/-)-alpha-Tocopherol; DL-all-rac-alpha-Tocopherol; Vitamin E
Type Certified Reference Material
Quantity 500 mg
Formula Weight 430.71
Grade Certified Reference Material
Show More Show Less