missing translation for 'onlineSavingsMsg'
Learn More

δ-Tocopherol, MilliporeSigma™ Supelco™

Catalog No. 111012719
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012719 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012719 Supplier Merck Emd Millipore Supplier No. 47784

δ-Tocopherol, a homolog of vitamin E, is a phytochemical with potent antioxidant activity. It has been investigated for its potential medical, biological, and physio-chemical applications.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products

Specifications

Packaging Ampule
CAS 119-13-1
Physical Form Neat
Linear Formula C27H46O2
MDL Number MFCD20486794
Synonym (+)-delta-Tocopherol
Type Food Additive Standard
Quantity 100 mg
Formula Weight 402.65
Grade Analytical Standard