missing translation for 'onlineSavingsMsg'
Learn More

TOC Standard, According to ISO/CEN EN1484, 25 mg/L±2 mg/L, MilliporeSigma™ Supelco™

Catalog No. p-7193485
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004585 250 mL
111004380 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111004585 Supplier Merck Emd Millipore Supplier No. 90326250MLF
Only null left

TOC standard can be used in the analysis of degradation by-products of acetaminophen (ACE) solution during electro-Fenton (EF) oxidation performed on carbon felt.

Specifications

Concentration 25 mg/L ±2 mg/L
Quantity 250 mL
Grade Analytical Standard