missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012194
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012194 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012194 Supplier Merck Emd Millipore Supplier No. PHR10791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Tobramycin is an aminoglycoside antibiotic effective in treating infections caused by Gram negative bacteria.

Specifications

CAS 32986-56-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C18H37N5O9
MDL Number MFCD00077885
Synonym Nebramycin Factor 6; O-[3-Amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)]-O-[2,6-diamino-2,3,6-trideoxy-alpha-D-ribohexopyranosyl-(1->4)]-2-deoxy-D-streptamine
Type Certified Reference Material
Quantity 1 g
Formula Weight 467.51
Grade Certified Reference Material