missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFSF13B, Rat anti-Human, Clone: 4.62, Abnova™
Rat Monoclonal Antibody
Supplier: Abnova Corporation MAB16092
Specifications
| TNFSF13B | |
| Monoclonal | |
| Unconjugated | |
| In PBS (10% glycerol, 0.02% sodium azide). | |
| BAFF/BLYS/CD257/DTL/TALL-1/TALL1/THANK/TNFSF20/ZTNF4 | |
| Rat | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Immunoprecipitation, Inhibition Assays | |
| 4.62 | |
| ELISA (as a detection antibody) Inhibition Assay Immunoprecipitation (1:200) The optimal working dilution should be determined by the end user. | |
| tumor necrosis factor (ligand) superfamily, member 13b | |
| TNFSF13B | |
| HA-tagged human TNFSF13B. | |
| RUO | |
| 10673 | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |