Learn More
Invitrogen™ Bovine TNF alpha ELISA Kit
Description
The Bovine Tumor Necrosis Factor alpha (Bov TNFα) ELISA quantitates Bov TNFα in bovine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Bov TNFα. Principle of the method The Bovine TNFα solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | Q06599 |
| Assay Range | 0.123 to 30 ng/mL |
| Assay Sensitivity | 0.1 ng/mL |
| Calibration Range | 0.1 ng/ml to 30 ng/ml |
| Description | TNF ng/mL to a (TNF) ELISA Kit, 96 Assays, 0.1 to 30 ng/mL Test Range, 5 hr. Test Time, 0.1 ng/mL Sensitivity, Cell Culture Supernatants, Plasma, Serum Sample, Cattle/Bovine Species, Colorimetric Detection Method, TNF Gene, Used for Protein Assays and Analysis, 2 to |
| Detectable Analytes | TNF alpha |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Product Type | ELISA |
| Show More |