missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa5116723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116723 Supplier Invitrogen™ Supplier No. PA5116723
Only null left

Rabbit Polyclonal Antibody

Positive Samples: U-251MG, HeLa, HepG2, A-549, Mouse liver, Mouse brain, Rat testis Immunogen sequence: KPPLYMGPEY IKYFNDKTID EELERDKRVT WIVEFFANWS NDCQSFAPIY ADLSLKYNCT GLNFGKVDVG RYTDVSTRYK VSTSPLTKQL PTLILFQGGK EAMRRPQIDK KGRAVSWTFS EENVIREFNL NELYQRAKKL SKAGDNIPEE QPVASTPTTV SDGENKKDK.

This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and a C-terminal ER-retention sequence. This protein is enriched on the mitochondria-associated-membrane of the ER via palmitoylation of two of its cytosolically exposed cysteines.

Specifications

Antigen TMX2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TMX2
Gene Accession No. Q5XIK2, Q9D710, Q9Y320
Gene Alias 2310042M24Rik; AA589631; Cell proliferation-inducing gene 26 protein; CGI-31; growth-inhibiting gene 11; My009; PDIA12; PIG26; protein disulfide isomerase family A, member 12; PSEC0045; thioredoxin domain containing 14; thioredoxin domain-containing protein 14; thioredoxin related transmembrane protein 2; thioredoxin-related transmembrane protein 2; thioredoxin-related transmembrane protein-like protein; TMX2; TXNDC14; Unknown (protein for MGC:134152); UNQ237/PRO270
Gene Symbols TMX2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 128-296 of human TMX2 (NP_0570431).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295701, 51075, 66958
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less