Learn More
Thermo Scientific™ TMTpro™ Zero Label Reagent
Description
Thermo Scientific™ TMTpro Zero Label Reagent serves as a 'light' tag for MS-level peptide quantitation for method optimization or use with the TMTpro 16plex Label Reagent Set.
Features of TMTpro Zero Label Reagent include:
- Economical – 'light' version may be used for optimization and method development of TMTpro-labeled samples
- Efficient – amine-reactive, NHS ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
- Compatible – optimized for use with high resolution Thermo Scientific™ MS/MS platforms, such as the Q Exactive and Orbitrap Fusion Tribrid instrument series, including the Orbitrap Eclipse Tribrid and Orbitrap Exploris 480 mass spectrometers with data analysis fully supported by Proteome Discoverer 2.3
TMTpro label reagents are the next generation of tandem mass tags, designed to increase the level of sample multiplexing without compromising on protein identification and quantitation. The TMTpro tag structure is similar to the TMT tag structure in that the tags remain isobaric and amine reactive, but differs in the longer spacer region and isobutyl proline mass reporter region. After MS/MS fragmentation, each TMTpro tag generates a unique reporter mass (i.e., TMTpro 126-134Da) in the low-mass region of the high-resolution MS/MS spectrum that is used for relative quantitation of protein expression levels. TMTpro Zero Label Reagent is a 'light' version of the TMTpro tag that may be used for method development or in conjunction with the set or subset of 'heavy' TMTpro reagents.
For high resolution analysis of TMTpro-labeled peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164942 or 164939). For the EASY-Spray source, the recommended LC column is the EASY-Spray C18 LC Column (Cat. No. ES802 or ES803).
These products are subject to a limited use label license.
Specifications
Specifications
| Description | TMTpro™ Zero Label Reagent, 5 x 0.5 mg |
| For Use With (Equipment) | Mass Spectrometer |
| Product Line | TMT |
| Content And Storage | Store at –20°C. |
| Label or Dye | Tandem Mass Tag™ (TMT) Labels |
| Shipping Condition | Dry Ice |
| Starting Material | Peptides |
| Final Product Type | Peptide |
| Detection Method | Mass Spectrometry |
| Workflow Step | Protein Labeling |
| Show More |
Frequently Asked Questions (FAQs)
The TMTpro Zero Label Reagent was designed to be used to optimize methods before multiplexed analysis of samples.
No, the TMT and TMTpro reagents cannot be mixed because the reporter ions are in the same region.
Yes, unused TMTpro label reagent that has been dissolved in anhydrous acetonitrile can be aliquoted into tubes, dried under vacuum at 25-30 degrees C, purged with nitrogen, and stored in a sealed bag with desiccant at -20 degrees C.
TMT labeling can be performed after reduction, alkylation, digestion, or prior to cleanup, if the sample is buffered at appropriate pH (8-8.5) in a suitable buffer without primary amines (e.g., Tris, glycine). TMT labeling can also be performed after peptide clean-up. Labeling of peptides after clean-up enables measuring and normalizing of the peptide samples for equal mixing.
Applying correction factors is required for more accurate relative protein quantitation as the isotopic impurity is variable among the tags, ranging from 5-10%.
For Research Use Only. Not for use in diagnostic procedures.