missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140180
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LHAATYTKKV LDARGSNSLP LLRSVLDKLS ANQQ Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 94%.
Acts as a regulator of the tubular endoplasmic reticulum (ER) network. Suppresses the RTN3/4-induced formation of the ER tubules. Positively regulates PERK-mediated and IRE1-mediated unfolded protein response signaling.Chemical Identifiers
| TMEM33 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| TMEM33 | |
| 1110006G02Rik; 1600019D15Rik; 2410089A21Rik; 2700052H22Rik; 5430406L04Rik; AA388285; AI314185; AI426615; db83; hypothetical protein LOC327214; Protein DB83; SHINC3; SHINC-3; Tmem33; Transmembrane protein 33; wu:fd16h10; wu:fi11f08; zgc:55931; zgc:77670 | |
| TMEM33 | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57088 | |
| 55161 | |
| Recombinant protein corresponding to Human TMEM33. Recombinant protein control fragment (Product #RP-109506). | |
| Antibody |
Specifications
| TMEM33 | |
| Polyclonal | |
| Unconjugated | |
| TMEM33 | |
| 1110006G02Rik; 1600019D15Rik; 2410089A21Rik; 2700052H22Rik; 5430406L04Rik; AA388285; AI314185; AI426615; db83; hypothetical protein LOC327214; Protein DB83; SHINC3; SHINC-3; Tmem33; Transmembrane protein 33; wu:fd16h10; wu:fi11f08; zgc:55931; zgc:77670 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57088 | |
| TMEM33 | |
| Recombinant protein corresponding to Human TMEM33. Recombinant protein control fragment (Product #RP-109506). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |