missing translation for 'onlineSavingsMsg'
Learn More

TMEM26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107293 Supplier Thermo Fisher Scientific Supplier No. PA5107293
Only null left

TMEM26 Polyclonal Antibody

TMEM26 Polyclonal Antibody for Western Blot

Specifications

Antigen TMEM26
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene TMEM26
Gene Accession No. Q6ZUK4
Gene Alias TMEM26; Transmembrane protein 26
Gene Symbols TMEM26
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human TMEM26.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219623
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less