missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM175 Recombinant Rabbit Monoclonal Antibody (5H18L65)

Catalog No. 703805
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
703805 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 703805 Supplier Invitrogen™ Supplier No. 703805
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Mouse, Rat, Bovine.

TMEM175 gene ontology annotations related to this gene include potassium channel activity.

Specifications

Antigen TMEM175
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 5H18L65
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene TMEM175
Gene Accession No. Q9BSA9
Gene Alias 3010001K23Rik; AI504381; endosomal/lysomomal potassium channel TMEM175; endosomal/lysosomal potassium channel TMEM175; hTMEM175; mTMEM175; RGD1309712; TMEM175; Transmembrane protein 175; transmembrane protein 175; endosomal/lysomomal potassium channel TMEM175
Gene Symbols TMEM175
Host Species Rabbit
Immunogen Peptides corresponding to human TMEM175 [1) aa12-aa29 2) aa282-aa299].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84286
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less