missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM107 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572774
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572774 Supplier Invitrogen™ Supplier No. PA572774
Only null left

Rabbit Polyclonal Antibody

TMEM107 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Gene ontology (GO) annotation includes embryonic digit morphogenesis; neural tube patterning; non-motile cilium assembly; protein localization to ciliary transition zone.

Specifications

Antigen TMEM107
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMEM107
Gene Accession No. Q6UX40
Gene Alias 1110004B13Rik; 2810049P21Rik; DC20; GRVS638; PRO1268; TMEM107; Transmembrane protein 107; UNQ638/PRO1268
Gene Symbols TMEM107
Host Species Rabbit
Immunogen TMEM107 antibody for a 19 amino acid peptide near the amino terminus of human TMEM107. The immunogen is within amino acids 20 - 70 of TMEM107.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84314
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less