missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEFF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150081
Description
TMEFF2 Polyclonal Antibody for Western Blot, ICC/IF, IHC
TMEFF2 may be a survival factor for hippocampal and mesencephalic neurons. The shedded form up-regulates cancer cell proliferation, probably by promoting ERK1/2 phosphorylation.Specifications
| TMEFF2 | |
| Polyclonal | |
| Unconjugated | |
| TMEFF2 | |
| 4832418D20Rik; 7630402F16Rik; cancer/testis antigen family 120, member 2; CT120.2; HPP1; Hyperplastic polyposis protein 1; MNCb-1026; TENB2; TMEFF2; tomoregulin; tomoregulin-2; TPEF; TR; TR-2; transmembrane protein TENB2; transmembrane protein with EGF like and two follistatin like domains 2; transmembrane protein with EGF-like and two follistatin-like domains; transmembrane protein with EGF-like and two follistatin-like domains 2; UNQ178/PRO204 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23671 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9UIK5 | |
| TMEFF2 | |
| Recombinant Tomoregulin (TR) (Thr52-Arg290). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |