missing translation for 'onlineSavingsMsg'
Learn More

Tizanidine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7137264
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25275G 5 g
T252725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25275G Supplier TCI America Supplier No. T25275G
Only null left

Chemical Identifiers

CAS 64461-82-1
Molecular Formula C9H9Cl2N5S
Molecular Weight (g/mol) 290.166
MDL Number MFCD00798231
InChI Key ZWUKMNZJRDGCTQ-UHFFFAOYSA-N
Synonym 5-Chloro-4-[(4,5-dihydro-1H-imidazol-2-yl)amino]-2,1,3-benzothiadiazole Hydrochloride
PubChem CID 114869
IUPAC Name 5-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine;hydrochloride
SMILES C1CN=C(N1)NC2=C(C=CC3=NSN=C32)Cl.Cl

Specifications

Melting Point 280°C
Color White-Yellow
Quantity 5 g
Formula Weight 290.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tizanidine Hydrochloride
RTECSNumber DK9910000
TSCA No