accessibility menu, dialog, popup

UserName

CAS RN 64461-82-1

IUPAC Name: hydrogen 5-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine chloride
Synonyms: hydrogen tizanidine chloride
Molecular Weight (g/mol): 290.17
Molecular Formula: C9H9Cl2N5S
InChi Key: ZWUKMNZJRDGCTQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=C(NC2=NCCN2)C2=NSN=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results