missing translation for 'onlineSavingsMsg'
Learn More

Titriplex™ II, Ethylenedinitrilotetraacetic acid, ACS, MilliporeSigma™

Catalog No. 361041999
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361041999 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361041999 Supplier Technidata Supplier No. 1.08417.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity ethylenedinitrilotetraacetic acid suitable for analytical applications.

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Density 600kg/m3
pH 2.5
Ignition Point ≥200°C
Packaging Plastic Bottle
Quantity 1 kg
Solubility Information 0.5g/L
Vapor Pressure ≥0.013 hPa (20°C)
Formula Weight 292.24
Percent Purity ACS, Reag. Ph
Grade ACS
Physical Form Fine Crystalline to Crystalline Powder
Assay 99.4 - 100.6%
Chemical Name or Material Ethylenedinitrilotetraacetic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category H319: Causes serious eye irritation.
Hazard Statement P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 200-449-4
RTECSNumber AH4025000