missing translation for 'onlineSavingsMsg'
Learn More

Titriplex™ II, For Analysis, Ethylenedinitrilotetraacetic acid, ACS, MilliporeSigma™

Catalog No. 361041998
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361041998 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361041998 Supplier Technidata Supplier No. 1.08417.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS reagent grade EDTA suitable for complexometric titration.

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color White
pH 2.5
Physical Form Fine Crystals or Crystalline Powder
Quantity 250 g
Assay Percent Range 99.4 - 100.6%
Percent Purity 99.7%
Grade ACS/Reagent Ph. Eur.
Ignition Point ≥200°C
Packaging Plastic bottle
Vapor Pressure ≤0.013 hPa (20°C)
Decomposition Information 220°C (Decomposition)
Density 0.86g/cm3 (20°C); Bulk Density: 600 kg/m3
Chemical Name or Material Ethylenedinitrilotetraacetic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category H319: Causes serious eye irritation.
Hazard Statement P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 200-449-4
RTECSNumber AH4025000
storageNote1 Storage temperature: No restrictions.