missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titration (TAN) Solvent, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Titration (TAN) Solvent |
| Color | Colorless |
| CAS Max % | 50, 49.5, 0.5 |
| CAS | 67-63-0, 7732-18-5 |
| Linear Formula | C7H8 |
| UN Number | UN1993 |
| Solubility Information | Soluble in water |
| Formula Weight | 92.14 |
| Grade | Certified |
| Identification | Passes Test |
| Show More |