missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titration Solvent, Fisher Chemical
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Specifications
| Chemical Name or Material | Titration Solvent |
| Color | Undesignated |
| Boiling Point | 80.0° to 81.0°C (20 drops); 110.1° to 111.1°C (95mL) |
| CAS | 67-63-0 |
| Assay Percent Range | 2-Propanol: 48.5 to 50.5%; Toluene: 49.0 to 51.0%; Water: 0.3 to 0.7% |
| MDL Number | MFCD00214201 |
| UN Number | 1993 |
| Density | 0.82g/cm³ |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Show More |