missing translation for 'onlineSavingsMsg'
Learn More

Titanyl Phthalocyanine (purified by sublimation), TCI America™

Catalog No. T2272200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2272200MG 200 mg
T22721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2272200MG Supplier TCI America Supplier No. T2272200MG
Only null left

Chemical Identifiers

CAS 26201-32-1
Molecular Formula C32H18N8OTi
Molecular Weight (g/mol) 578.418
MDL Number MFCD00145414
InChI Key DOEAHNYJXVICHS-UHFFFAOYSA-N
Synonym TiOPc, Oxo(phthalocyaninato)titanium
PubChem CID 131664299
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2[N-]3.O.[Ti+2]

Specifications

Melting Point 395°C
Color Blue-Purple
Quantity 200 mg
Formula Weight 576.40
Physical Form Crystalline Powder
Chemical Name or Material Titanyl Phthalocyanine (purified by sublimation)