accessibility menu, dialog, popup

UserName

CAS RN 26201-32-1

OTiNNNNNNNN

CAS RN 26201-32-1

IUPAC Name: 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),3(40),4,6,8,10,12(39),13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-2,37-diide; oxotitaniumbis(ylium)
Synonyms: 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),3(40),4,6,8,10,12(39),13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-2,37-diide; oxotitaniumbis(ylium)
Molecular Weight (g/mol): 576.4
Molecular Formula: C32H16N8OTi
InChi Key: SJHHDDDGXWOYOE-UHFFFAOYSA-N
SMILES: O=[Ti++].[N-]1C2=C3C=CC=CC3=C1N=C1N=C(N=C3N=C(N=C4N=C([N-]2)C2=CC=CC=C42)C2=CC=CC=C32)C2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results