missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, catalyst support

Anatase, Typical properties = S.A. > 150m2/g, median pore diameter 140Å, total pore volume 0.38cc/g, packing density 0.90kg/L, contains 1% S (as SO4) | CAS: 1317-70-0 | O2Ti | 79.87 g/mol

Supplier:  Thermo Scientific Chemicals 044429A1

 View more versions of this product

Catalog No. AA44429A1


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Titanium(IV) oxide, catalyst support
1317-70-0
1 kg
MFCD00011269,MFCD00210650
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.9
1/8 in. Pellets
Pellets
O2Ti
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
O=[Ti]=O
79.87
CHEBI:32234

Chemical Identifiers

1317-70-0
79.87
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269,MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
CHEBI:32234
O=[Ti]=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-280-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only