missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, catalyst support

Catalog No. p-4869448
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4442930 250 g
AA44429A1 1 kg
AA44429A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4442930 Supplier Thermo Scientific Chemicals Supplier No. 04442930

Anatase, Typical properties = S.A. > 150m2/g, median pore diameter 140Å, total pore volume 0.38cc/g, packing density 0.90kg/L, contains 1% S (as SO4) | CAS: 1317-70-0 | O2Ti | 79.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1317-70-0
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269,MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Physical Form Pellets
Quantity 250 g
Formula Weight 79.9
Chemical Name or Material Titanium(IV) oxide, 1/8 in. Pellets
EINECSNumber 215-280-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only