missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) ethoxide

Catalog No. AC312542500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC312542500 250 g Glass bottle
AC312540500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC312542500 Supplier Thermo Scientific Chemicals Supplier No. 312542500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3087-36-3 | C8H20O4Ti | 228.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Chemical Identifiers

CAS 3087-36-3 , 67-63-0
Molecular Formula C8H20O4Ti
Molecular Weight (g/mol) 228.15
MDL Number MFCD00009071
InChI Key JMXKSZRRTHPKDL-UHFFFAOYSA-N
Synonym titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
PubChem CID 76524
IUPAC Name ethanolate;titanium(4+)
SMILES CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]

Specifications

Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 150.0°C to 152.0°C (10.0 mmHg)
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 85% min
Packaging Glass bottle
Refractive Index 1.5033 to 1.5053
Linear Formula Ti(OC2H5)4
Quantity 250 g
Beilstein 01,III,1338
Specific Gravity 1.08
Solubility Information Solubility in water: hydrolysis.
Formula Weight 228.15
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material Titanium(IV) ethoxide, Contains 5-15% Isopropanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-410-8

RUO – Research Use Only