missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) chloride, 1M soln. in dichloromethane

Catalog No. AAH31830AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31830AE 100 mL
AAH31830AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31830AE Supplier Thermo Scientific Chemicals Supplier No. H31830AE

CAS: 7550-45-0 | Cl4Ti | 189.667 g/mol

Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Activates pyrrolidines for improved conversion, via a modified Bouveault reaction, to the corresponding α,α-dimethylamines

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Chemical Identifiers

CAS 7550-45-0
Molecular Formula Cl4Ti
Molecular Weight (g/mol) 189.667
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Quantity 100 mL
Linear Formula TiCl4
UN Number UN2922
Solubility Information Reacts with water.
Formula Weight 189.69
Concentration or Composition (by Analyte or Components) 1M soln. in dichloromethane
Sensitivity Moisture sensitive; May form a precipitate
Density 1.365
Chemical Name or Material Titanium(IV) chloride
Hazard Category H314-H331-H335-H336-H351
Hazard Statement GHS H Statement
H314-H351
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P271-P281-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only