missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) 2-ethylhexyloxide, 95%

Catalog No. p-7080292
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA7710722 100 g
AA7710736 500 g
AA77107A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA7710722 Supplier Thermo Scientific Chemicals Supplier No. 07710722
Only null left

CAS: 1070-10-6 | C32H68O4Ti | 564.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1070-10-6
Molecular Formula C32H68O4Ti
Molecular Weight (g/mol) 564.76
MDL Number MFCD00015264,MFCD00015264
InChI Key KTXWGMUMDPYXNN-UHFFFAOYNA-N
Synonym tyzor tot, 2-ethylhexyl orthotitanate, tetra 2-ethylhexyl titanate, tetra-2-ethylhexyl titanate, tetrakis 2-ethylhexyl titanate, titanic iv acid, tetrakis 2-ethylhexyl ester, 1-hexanol, titanium 4+ salt
PubChem CID 54603245
IUPAC Name titanium(4+) tetrakis(2-ethylhexan-1-olate)
SMILES [Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-]

Specifications

Boiling Point 194°C (0.25mmHg)
Melting Point <-25°C
Color Yellow
Physical Form Liquid
Quantity 100 g
Linear Formula Ti[OCH2CH(C2H5)(CH2)3CH3]4
Beilstein 3692023
Flash Point 71°C (160°F)
Formula Weight 564.79
Percent Purity 95%
Refractive Index 1.482
Sensitivity Moisture sensitive
Density 0.926 g/mL
Chemical Name or Material Titanium (IV) 2-ethylhexyloxide
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 213-969-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only