missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(III) fluoride
Description
It is used as a laboratory chemicals & for manufacture of chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 10 g |
| UN Number | UN3260 |
| Solubility Information | Sparingly soluble. It hydrolyses in water. |
| Formula Weight | 104.9 |
| Odor | Pungent, makes eyes water |
| Sensitivity | Air and moisture sensitive |
| Density | 3.400 g/mL |
| Chemical Name or Material | Titanium(III) fluoride |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318-H302-H312-H332 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 236-732-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only