missing translation for 'onlineSavingsMsg'
Learn More

Medisca Titanium Dioxide, USP

Catalog No. 14185532
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185532 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14185532 Supplier Medisca Supplier No. 165801

Titanium Dioxide USP is a white, odorless powder.

Titanium Dioxide USP is a white, odorless powder.

Specifications

Chemical Name or Material Titanium Dioxide, USP
Packaging Jar, 4L, White, HDPE, Round, 110-400,TE Pano Neck
CAS 13463-67-7
Type Drug Standard
Molecular Weight (g/mol) 79.87
Quantity 2.5 kg
Percent Purity 99 to 100.5 %
Grade USP