Learn More
Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol
CAS: 17927-72-9 | C16H28O6Ti
$64.96 - $178.30
Chemical Identifiers
| CAS | 17927-72-9 |
|---|---|
| Molecular Formula | C16H28O6Ti |
| MDL Number | MFCD00000031 |
| Synonym | Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV) |
Description
Titanium(diisopropoxide) bis(2,4-pentanedionate) is used as catalyst, used in sol-gel, coating, primer and crosslinking.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 17927-72-9 | |
| C16H28O6Ti | |
| MFCD00000031 | |
| Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV) |
Specifications
| Brown | |
| Liquid | |
| C16H28O6Ti | |
| UN1219 | |
| Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV) | |
| 364.3 | |
| Alcohol-like | |
| Moisture sensitive | |
| Titanium (diisopropoxide) bis (2,4-pentanedionate) |
| 17927-72-9 | |
| 100 g | |
| MFCD00000031 | |
| 18°C (61°F) | |
| Hydrolyze with water. | |
| 75% in isopropanol | |
| 1.494 | |
| 1.010 g/mL |
Safety and Handling
GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H226-H319-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
EINECSNumber : 241-866-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only