missing translation for 'onlineSavingsMsg'
Learn More

Titanium, AAS standard solution, Specpure™ Ti 1000μg/mL

$77.51 - $77.51

Chemical Identifiers

MDL Number MFCD00011264
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35771AE
View Documents
Thermo Scientific Chemicals
035771AE
100 mL
Each for $77.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00011264

Specifications

Liquid
MFCD00011264
100 mL
Titanium AAS standard solution, Ti 1000μg/mL
Ti in 5% HNO3/tr. HF
UN3264
Specpure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only