Learn More
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes
For biopsy tissue samples, these cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties
$926.22
Specifications
| Closure Included | Yes |
|---|---|
| Quantity | 1000/Cs. |
| Pore Style | Small Squares |
| Closure Type | Separate Lid, Back Hinge |
| PrintMate Compatible | No |
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|
B851925BE
|
Epredia™
B851925BE |
Beige |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925BL
|
Epredia™
B851925BL |
Blue |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925GN
|
Epredia™
B851925GN |
Green |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925GY
|
Epredia™
B851925GY |
Gray |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925LA
|
Epredia™
B851925LA |
Lavender |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925LG
|
Epredia™
B851925LG |
Light Green |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925LI
|
Epredia™
B851925LI |
Lilac |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925OR
|
Epredia™
B851925OR |
Orange |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925TN
|
Epredia™
B851925TN |
Tan |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925WH
|
Epredia™
B851925WH |
White |
Case of 1000 Each for $926.22
|
|
|||||
|
B851925YW
|
Epredia™
B851925YW |
Yellow |
Case of 1000 Each for $926.22
|
|
|||||
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
| Yes | |
| Small Squares | |
| No |
| 1000/Cs. | |
| Separate Lid, Back Hinge | |
| TissueLoc Cassette IV with Color-Matched Lids |