missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tissue Detail Set
Description
Includes
- Squamous epithelium from human mouth, w.m.
- Columnar epithelium from human gall bladder, t.s.
- Ciliated epithelium from human trachea, t.s.
- Skin, human
- Human scalp, l.s.
- Developing nail, human embryo, l.s.
- Hyaline cartilage, human, t.s.
- Elastic cartilage, ear of pig, t.s.
- Developing cartilaginous bone, human fetus, l.s.
- Compact bone, c.s. and l.s.
- Striated muscle, human, l.s., staining of striations
- Striated muscle, human, t.s.
- Smooth muscle, human, t.s. and l.s.
- White fibrous tissue, human tendon, l.s.
- Adipose tissue, human, t.s.
Contains 15 of the most popular tissue slides.
Specifications
Specifications
| Type | Tissues Detail Set |