missing translation for 'onlineSavingsMsg'
Learn More

TCI America™ Tissue-Clearing Reagent CUBIC-X2

CUBIC-X2 is an optional addition to the CUBIC clearing process, and is used for RI matching with expanded tissues. Used in combination with CUBIC-X1 (T3866).

$61.25 - $198.19

Specifications

For Use With (Equipment) Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Product Line CUBIC
For Use With (Application) Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
T386725ML
View Documents Promotion Details
TCI America™
T386725ML
25 mL
Each for $61.25
Only null left
 
T3867100ML
View Documents Promotion Details
TCI America™
T3867100ML
100 mL
Each for $198.19
Only null left
 
Description

Description

  • The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
  • CUBIC-X2 is an optional addition to the CUBIC clearing protocol, and is used for RI matching on previously swollen/expanded samples.

Note: Both CUBIC-X2 (T3867) and CUBIC-X1 (T3866) are required for the CUBIC clearing process with expansion/swelling.

Specifications

Specifications

Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
CUBIC
Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Liquid
Videos
SDS
Documents

Documents