Learn More
TCI America™ Tissue-Clearing Reagent CUBIC-X2
CUBIC-X2 is an optional addition to the CUBIC clearing process, and is used for RI matching with expanded tissues. Used in combination with CUBIC-X1 (T3866).
Supplier: TCI America™ T3867100ML
Description
- The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
- CUBIC-X2 is an optional addition to the CUBIC clearing protocol, and is used for RI matching on previously swollen/expanded samples.
Note: Both CUBIC-X2 (T3867) and CUBIC-X1 (T3866) are required for the CUBIC clearing process with expansion/swelling.
Chemical Identifiers
| Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device | |
| CUBIC | |
| Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications | |
| Liquid |
Specifications
| 100 mL | |
| CUBIC | |
| Liquid |
| Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device | |
| Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications |