missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCI America™ Tissue-Clearing Reagent CUBIC-R+(N)
CUBIC-R+(N) is a more economical option for RI matching compared to CUBIC-R+(M) if fluorescent signal detection is not critical.
$93.17 - $666.86
Specifications
| For Use With (Equipment) | Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device |
|---|---|
| Product Line | CUBIC |
| For Use With (Application) | Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications |
| Form | Liquid |
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|
T398325ML
|
TCI America™
T398325ML |
25 mL |
Each for $93.17
|
|
|||||
|
T3983100ML
|
TCI America™
T3983100ML |
100 mL |
Each for $221.06
|
|
|||||
|
T3983500ML
|
TCI America™
T3983500ML |
500 mL |
Each for $666.86
|
|
|||||
Description
- The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
- CUBIC-R+(M) (T3741) is used for RI matching while CUBIC-L (T3740) is used for delipidation.
Note: Both T3893 (or T3741) and T3740 are required for the CUBIC clearing process.
Specifications
| Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device | |
| CUBIC | |
| Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications | |
| Liquid |