missing translation for 'onlineSavingsMsg'
Learn More

TCI America™ Tissue-Clearing Reagent CUBIC-R+(N)

Catalog No. t3983500ml
Change view
Click to view available options
Capacity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
T3983500ML 500 mL
T398325ML 25 mL
T3983100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T3983500ML Supplier TCI America™ Supplier No. T3983500ML
Only null left

CUBIC-R+(N) is a more economical option for RI matching compared to CUBIC-R+(M) if fluorescent signal detection is not critical.

  • The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
  • CUBIC-R+(M) (T3741) is used for RI matching while CUBIC-L (T3740) is used for delipidation.

Note: Both T3893 (or T3741) and T3740 are required for the CUBIC clearing process.

Specifications

Capacity 500 mL
For Use With (Equipment) Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Product Line CUBIC
For Use With (Application) Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Form Liquid