missing translation for 'onlineSavingsMsg'
Learn More

TISAB IV Solution, Reagents

Catalog No. 01333435
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333435 1 L
01333436 10 L
01333434 20 L
01333433 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01333435 Supplier Labchem Inc Supplier No. CT1300001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TISAB IV Solution, Reagent, use for Water analysis (Fluoride). Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material TISAB IV Solution
CAS 7732-18-5, 77-86-1, 6106-24-7
Boiling Point 100°C
Color Colorless
For Use With (Equipment) Fluoride Meter
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.1
Sterility Non-sterile
Synonym Total Ionic Strength Adjustment Buffer
Grade Reagent
Show More Show Less