missing translation for 'onlineSavingsMsg'
Learn More

Tiron Monohydrate 98.0+%, TCI America™

Catalog No. p-7126292
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D056725G 25 g
D0567250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D056725G Supplier TCI America Supplier No. D056725G
Only null left

Chemical Identifiers

CAS 149-45-1
Molecular Formula C6H10Na2O8S2
Molecular Weight (g/mol) 320.238
MDL Number MFCD00007473
InChI Key HEOKHLCODUWALT-UHFFFAOYSA-N
Synonym tiron, tiferron, sodium catechol sulfate, disodium 4,5-dihydroxybenzene-1,3-disulfonate, 3,5-disulfocatechol disodium salt, sodium pyrocatechol-3,5-disulfonate, disodium pyrocatechol-3,5-disulfonate, sodium 1,2-dihydroxybenzenedisulfonate, 1,2-dihydroxybenzene-3,5-disulfonic acid disodium salt, 1,3-benzenedisulfonic acid, 4,5-dihydroxy-, disodium salt
PubChem CID 131674010
IUPAC Name 4,5-dihydroxybenzene-1,3-disulfonic acid;molecular hydrogen;sodium
SMILES [HH].[HH].C1=C(C=C(C(=C1S(=O)(=O)O)O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Color White
Quantity 25 g
Formula Weight 314.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tiron Monohydrate
EINECSNumber (3)-3848
RTECSNumber CZ9263000
TSCA Yes