missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate, Heptahydrate, Granular, USP, 99.5-104.5%, Spectrum™ Chemical

Catalog No. 18602726
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602726 50 kg Poly Bottle
18602723 500 g Poly Bottle
18602724 2.5 kg Poly Bottle
18602725 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602726 Supplier Spectrum Chemical Mfg Cor Supplier No. FE11050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FE110, 7782-63-0, FeSO4·7H2O

Spectrum™ Chemical Ferrous Sulfate, Heptahydrate, Granular, USP is used to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
IUPAC Name λ2-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Physical Form Granules
CAS Max % 100%
Quantity 50 kg
Packaging Poly Bottle
Percent Purity 99.5 to 104.5%
Grade USP
Chemical Name or Material Ferrous Sulfate, Heptahydrate, Granular