missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiotropium Bromide 98.0+%, TCI America™
Supplier: TCI America T3032200MG
Specifications
| Tiotropium Bromide | |
| 230°C | |
| C19H22BrNO4S2 | |
| 200 mg | |
| C[N+]1(C2CC(CC1C3C2O3)OC(=O)C(C4=CC=CS4)(C5=CC=CS5)O)C.[Br-] | |
| 5487426 | |
| ≥98.0% (HPLC,T) |
| 136310-93-5 | |
| White-Yellow | |
| MFCD00867027 | |
| DQHNAVOVODVIMG-RGECMCKFSA-M | |
| 472.412 | |
| 472.41 | |
| Crystalline Powder |
Chemical Identifiers
| 136310-93-5 | |
| 472.412 | |
| DQHNAVOVODVIMG-RGECMCKFSA-M | |
| C[N+]1(C2CC(CC1C3C2O3)OC(=O)C(C4=CC=CS4)(C5=CC=CS5)O)C.[Br-] |
| C19H22BrNO4S2 | |
| MFCD00867027 | |
| 5487426 |
Safety and Handling
TSCA : No